2-Fluoro-3-(trifluoromethyl)benzyl alcohol 97%
Supplier: Apollo Scientific
Warning
PC4375E-10G PC4375E-1G PC4375E-5G PC4375E-25G
APOSPC4375E-10GEA 121 EUR
APOSPC4375E-10G APOSPC4375E-1G APOSPC4375E-5G APOSPC4375E-25G
2-Fluoro-3-(trifluoromethyl)benzyl alcohol 97%
2-Fluoro-3-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆F₄O
MW: 194.13 g/mol
Boiling Pt: 211 °C
Melting Pt: 24…26 °C
Density: 1.397 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00061166
CAS Number: 207981-45-1