2-(Difluoromethoxy)aniline 98%
Supplier: Apollo Scientific
Warning
PC4376-100G PC4376-25G
APOSPC4376-100GEA 0 EUR
APOSPC4376-100G APOSPC4376-25G
2-(Difluoromethoxy)aniline 98%
2-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 205 °C
Density: 1.272 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00190104
CAS Number: 22236-04-0