2-Fluoro-4-(trifluoromethyl)benzyl bromide 98%
Supplier: Apollo Scientific
Danger
PC4376F-25G PC4376F-5G PC4376F-100G
APOSPC4376F-25GEA 217 EUR
APOSPC4376F-25G APOSPC4376F-5G APOSPC4376F-100G
2-Fluoro-4-(trifluoromethyl)benzyl bromide 98%
2-Fluoro-4-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Boiling Pt: 197
Melting Pt: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00061173
CAS Number: 239087-07-1
UN: 1759
ADR: 8 III