3-Fluoro-5-(trifluoromethyl)benzyl bromide 98%
Supplier: Apollo Scientific
Danger
PC4376I-10G PC4376I-5G PC4376I-25G
APOSPC4376I-10GEA 126 EUR
APOSPC4376I-10G APOSPC4376I-5G APOSPC4376I-25G
3-Fluoro-5-(trifluoromethyl)benzyl bromide 98%
3-Fluoro-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrF₄
MW: 257.03 g/mol
Boiling Pt: 43 °C/1mmHg
Storage Temperature: Ambient
MDL Number: MFCD00061175
CAS Number: 239087-09-3
UN: 1760
ADR: 8 III