2-Fluoro-5-(trifluoromethyl)phenol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Fluor-2-hydroxy-4-(trifluormethyl)benzol, 2-Fluoro-5-trifluoromethylphenol
PC4376O-25G PC4376O-5G
APOSPC4376O-25GEA 37.7 EUR
APOSPC4376O-25G APOSPC4376O-5G
2-Fluoro-5-(trifluoromethyl)phenol 98%
2-Fluoro-5-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 146 °C
Density: 1.436 g/cm³ (25 °C)
Flash Pt: 57 °C
Storage Temperature: Ambient
MDL Number: MFCD00061286
CAS Number: 141483-15-0
UN: 2920
ADR: 8+3,II