4-Fluoro-3-(trifluoromethyl)phenol 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-Fluoro-5-hydroxybenzotrifluoride, 4-Fluoro-3-trifluoromethylphenol
PC4376R-100G PC4376R-25G PC4376R-5G
APOSPC4376R-100GEA 136 EUR
APOSPC4376R-100G APOSPC4376R-25G APOSPC4376R-5G
4-Fluoro-3-(trifluoromethyl)phenol 98%
4-Fluoro-3-(trifluoromethyl)phenol
Formula: C₇H₄F₄O
MW: 180.1 g/mol
Boiling Pt: 84…86 °C/15mmHg
Melting Pt: 17 °C
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00043875
CAS Number: 61721-07-1
EINECS: 432-560-0
UN: 3265
ADR: 8,II