2-Fluoro-6-(trifluoromethyl)phenylacetic acid 98%
Supplier: Apollo Scientific
Warning
PC4376V-10G PC4376V-5G PC4376V-1G
APOSPC4376V-10GEA 250 EUR
APOSPC4376V-10G APOSPC4376V-5G APOSPC4376V-1G
2-Fluoro-6-(trifluoromethyl)phenylacetic acid 98%
2-Fluoro-6-(trifluoromethyl)phenylacetic acid
Formula: C₉H₆F₄O₂
MW: 222.14 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00061191
CAS Number: 179946-32-8