2-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2-(2-Fluoro-3-(trifluoromethyl)phenyl)acetonitrile
PC4377E-10G PC4377E-1G PC4377E-5G
APOSPC4377E-10GEA 149 EUR
APOSPC4377E-10G APOSPC4377E-1G APOSPC4377E-5G
2-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%
2-Fluoro-3-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
Boiling Pt: 237 °C
Density: 1.34 g/cm³ (25 °C)
Flash Pt: 106 °C
Storage Temperature: Ambient
MDL Number: MFCD00061178
CAS Number: 239087-10-6
UN: 3439
ADR: 6.1,III