2-Fluoro-4-(trifluoromethyl)phenylacetonitrile 94%
Supplier: Apollo Scientific
Danger
PC4377F-10G PC4377F-5G PC4377F-1G
APOSPC4377F-10GEA 329 EUR
APOSPC4377F-10G APOSPC4377F-5G APOSPC4377F-1G
2-Fluoro-4-(trifluoromethyl)phenylacetonitrile 94%
2-Fluoro-4-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
Storage Temperature: Ambient
MDL Number: MFCD00061179
CAS Number: 239087-11-7
UN: 3276
ADR: 6.1 II