2-Fluoro-6-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
PC4377H-5G PC4377H-1G PC4377H-25G
APOSPC4377H-5GEA 92.8 EUR
APOSPC4377H-5G APOSPC4377H-1G APOSPC4377H-25G
2-Fluoro-6-(trifluoromethyl)phenylacetonitrile 98%
2-Fluoro-6-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
Boiling Pt: 229 °C
Density: 1.363 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061184
CAS Number: 179946-34-0