3-Fluoro-5-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Warning
PC4377J-10G PC4377J-5G PC4377J-1G
APOSPC4377J-10GEA 119 EUR
APOSPC4377J-10G APOSPC4377J-5G APOSPC4377J-1G
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile 98%
3-Fluoro-5-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
MW: 203.1400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061181
CAS Number: 239087-12-8
UN: 3276
ADR: 6.1 III