4-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-fluoro-3-(trifluoromethyl)phenylacetonitrile
PC4377M-10G PC4377M-1G PC4377M-5G
APOSPC4377M-10GEA 52.2 EUR
APOSPC4377M-10G APOSPC4377M-1G APOSPC4377M-5G
4-Fluoro-3-(trifluoromethyl)phenylacetonitrile 98%
4-Fluoro-3-(trifluoromethyl)phenylacetonitrile
Formula: C₉H₅F₄N
MW: 203.14 g/mol
Boiling Pt: 93 °C/10mmHg
Storage Temperature: Ambient
MDL Number: MFCD00061183
CAS Number: 220239-65-6
UN: 3276
ADR: 6.1 III