2'-Fluoro-3'-(trifluoromethyl)propiophenone 97%
Supplier: Apollo Scientific
Warning
PC4377P-25G PC4377P-5G
APOSPC4377P-25GEA 139 EUR
APOSPC4377P-25G APOSPC4377P-5G
2'-Fluoro-3'-(trifluoromethyl)propiophenone 97%
2'-Fluoro-3'-(trifluoromethyl)propiophenone
Formula: C₁₀H₈F₄O
MW: 220.17 g/mol
Boiling Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00061192
CAS Number: 207986-23-0