5-Fluoro-DL-tryptophan
Supplier: Apollo Scientific
Warning
Synonyms: 2-Amino-3-(5-fluoro-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-F)-OH
PC4377Z-1G PC4377Z-250MG
APOSPC4377Z-1GEA 111 EUR
APOSPC4377Z-1G APOSPC4377Z-250MG
5-Fluoro-DL-tryptophan
5-Fluoro-DL-tryptophan
Formula: C₁₁H₁₁FN₂O₂
Melting Pt: 265 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00005649
CAS Number: 154-08-5
EINECS: 205-822-5