Gallium trifluoride 99%
Supplier: Apollo Scientific
Danger
PC4387L-1G PC4387L-5G
APOSPC4387L-1GEA 198 EUR
APOSPC4387L-1G APOSPC4387L-5G
Gallium trifluoride 99%
Gallium trifluoride
Formula: F₃Ga
MW: 126.72 g/mol
Boiling Pt: 1000 °C
Melting Pt: 800 °C
Density: 4.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011021
CAS Number: 7783-51-9
EINECS: 232-004-5
UN: 3260
ADR: 8,III