2,2,3,3,4,4,4-Heptafluorobutylamine 97%

Supplier: Apollo Scientific
Danger

APOSPC4396-1GEA 21.9 EUR
APOSPC4396-1G
2,2,3,3,4,4,4-Heptafluorobutylamine 97%
2,2,3,3,4,4,4-Heptafluorobutylamine
Formula: C₄H₄F₇N
MW: 199,07 g/mol
Boiling Pt: 70…71 °C
Density: 1,493 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
EINECS: 206-780-0
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR