2,6-Difluoro-DL-phenylalanine 97%
Supplier: Apollo Scientific
Warning
PC4398-1G PC4398-250MG PC4398-5G
APOSPC4398-1GEA 87 EUR
APOSPC4398-1G APOSPC4398-250MG APOSPC4398-5G
2,6-Difluoro-DL-phenylalanine 97%
2,6-Difluoro-DL-phenylalanine
Formula: C₉H₉F₂NO₂
Melting Pt: 256…257 °C
Storage Temperature: Ambient
MDL Number: MFCD00061304
CAS Number: 32133-39-4