4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Fluoro-2-(trifluoromethyl)benzene-1-sulphonyl chloride
PC4401-1G PC4401-25G PC4401-5G
APOSPC4401-1GEA 21.9 EUR
APOSPC4401-1G APOSPC4401-25G APOSPC4401-5G
4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride 98%
4-Fluoro-2-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃ClF₄O₂S
MW: 262.61 g/mol
Boiling Pt: 76 °C/0.5mmHg
Melting Pt: 48…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01091000
CAS Number: 176225-09-5
UN: 3261
ADR: 8 III