3-Methyl-5-(trifluoromethyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methyl-5-(trifluoromethyl)pyrazole
PC4439-1G PC4439-25G PC4439-5G
APOSPC4439-1GEA 21.9 EUR
APOSPC4439-1G APOSPC4439-25G APOSPC4439-5G
3-Methyl-5-(trifluoromethyl)-1H-pyrazole
3-Methyl-5-(trifluoromethyl)-1H-pyrazole
Formula: C₅H₅F₃N₂
MW: 150.104 g/mol
Boiling Pt: 117 °C/15mmHg
Melting Pt: 89…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00156058
CAS Number: 10010-93-2