2-(5,6-Difluoro-1H-benzoimidazol-2-yl)ethylamine
Supplier: Apollo Scientific
PC446007-1G PC446007-500MG
APOSPC446007-1GEA 412 EUR
APOSPC446007-1G APOSPC446007-500MG
2-(5,6-Difluoro-1H-benzoimidazol-2-yl)ethylamine
2-(5,6-Difluoro-1H-benzoimidazol-2-yl)ethylamine
Formula: C₉H₉F₂N₃
MW: 197.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568201
CAS Number: 1053656-20-4