4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine
Supplier: Apollo Scientific
Synonyms: 2-Trifluoromethyl-pyrimidine-4,6-diol, 2-(Trifluoromethyl)pyrimidine-4,6-diol
PC446044-1G
APOSPC446044-1GEA 166 EUR
APOSPC446044-1G
4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine
4,6-Dihydroxy-2-(trifluoromethyl)pyrimidine
Formula: C₅H₃F₃N₂O₂
MW: 180.09 g/mol
Melting Pt: 254…256 °C
Storage Temperature: Ambient
MDL Number: MFCD02678234
CAS Number: 672-47-9
UN: 2811
ADR: 6.1,III