(2-Trifluoromethoxyphenyl)hydrazine hydrochloride
Supplier: Apollo Scientific
Synonyms: (2-(Trifluoromethoxy)phenyl)hydrazine hydrochloride
PC446051-1G PC446051-5G PC446051-10G
APOSPC446051-1GEA 102 EUR
APOSPC446051-1G APOSPC446051-5G APOSPC446051-10G
(2-Trifluoromethoxyphenyl)hydrazine hydrochloride
(2-Trifluoromethoxyphenyl)hydrazine hydrochloride
Formula: C₇H₈ClF₃N₂O
MW: 228.6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782063
CAS Number: 133115-76-1