4-Trifluoromethyl-L-phenylalanine 95%
Supplier: Apollo Scientific
Synonyms: H-Phe(4-CF3)-OH, 4-(Trifluoromethyl)-L-phenylalanine
PC4463-10G PC4463-1G PC4463-5G
APOSPC4463-10GEA 800 EUR
APOSPC4463-10G APOSPC4463-1G APOSPC4463-5G
4-Trifluoromethyl-L-phenylalanine 95%
4-Trifluoromethyl-L-phenylalanine
Formula: C₁₀H₁₀F₃NO₂
MW: 233.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00044946
CAS Number: 114926-38-4