1-[3-(Trifluoromethyl)phenyl]propan-1-ol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-(Trifluoromethyl)phenyl)propan-1-ol
PC4465-1G PC4465-5G
APOSPC4465-1GEA 72.7 EUR
APOSPC4465-1G APOSPC4465-5G
1-[3-(Trifluoromethyl)phenyl]propan-1-ol 97%
1-[3-(Trifluoromethyl)phenyl]propan-1-ol
Formula: C₁₀H₁₁F₃O
MW: 204.19 g/mol
Boiling Pt: 88…91 °C/5mmHg
Density: 1.197 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00046668
CAS Number: 618-97-3