Methyl-3-(trifluoromethyl)benzoate 99+%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-(trifluoromethyl)benzoate
PC4482-100G PC4482-25G PC4482-500G
APOSPC4482-100GEA 43.5 EUR
APOSPC4482-100G APOSPC4482-25G APOSPC4482-500G
Methyl-3-(trifluoromethyl)benzoate 99+%
Methyl 3-(trifluoromethyl)benzoate
Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
Boiling Pt: 80…82 °C/15mmHg
Density: 1.295 g/cm³ (25 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00043543
CAS Number: 2557-13-3
UN: 3272
ADR: 3,III