2-Bromo-5-(trifluoromethyl)benzyl alcohol
Supplier: Apollo Scientific
Warning
Synonyms: 2-Bromo-5-(trifluoromethyl)benzyl Alcohol
PC4491-100G PC4491-25G PC4491-5G
APOSPC4491-100GEA 319 EUR
APOSPC4491-100G APOSPC4491-25G APOSPC4491-5G
2-Bromo-5-(trifluoromethyl)benzyl alcohol
2-Bromo-5-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆BrF₃O
MW: 255.319 g/mol
Melting Pt: 66…67 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04973761
CAS Number: 869725-53-1