6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione 97%
Supplier: Apollo Scientific
Warning
Synonyms: 6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyloctane-3,5-dione, 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione, HFOD
PC4496-100G PC4496-25G PC4496-5G
APOSPC4496-100GEA 466 EUR
APOSPC4496-100G APOSPC4496-25G APOSPC4496-5G
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione 97%
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione
Formula: C₁₀H₁₁F₇O₂
MW: 296.18 g/mol
Boiling Pt: 56…57 °C/10mmHg
Density: 1.272 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000435
CAS Number: 17587-22-3
EINECS: 241-556-6
UN: 1224
ADR: 3,III