2'-Fluoro-2'-deoxyadenosine
Supplier: Apollo Scientific
Warning
Synonyms: 2′-Deoxy-2′-fluoroadenosine, (2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)tetrahydrofuran-3-ol, 2'-Deoxy-2'-fluoroadenosine
PC450034-1G PC450034-500MG PC450034-5G PC450034-250MG
APOSPC450034-1GEA 102 EUR
APOSPC450034-1G APOSPC450034-500MG APOSPC450034-5G APOSPC450034-250MG
2'-Fluoro-2'-deoxyadenosine
2'-Fluoro-2'-deoxyadenosine
Formula: C₁₀H₁₂FN₅O₃
MW: 269.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09750859
CAS Number: 64183-27-3