2,4-Dimethoxy-1-fluorobenzene 97%
Supplier: Apollo Scientific
Synonyms: 1-Fluoro-2,4-dimethoxybenzene
PC450147-10G PC450147-1G PC450147-5G PC450147-25G
APOSPC450147-10GEA 0 EUR
APOSPC450147-10G APOSPC450147-1G APOSPC450147-5G APOSPC450147-25G
2,4-Dimethoxy-1-fluorobenzene 97%
2,4-Dimethoxy-1-fluorobenzene
Formula: C₈H₉FO₂
MW: 156.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042265
CAS Number: 17715-70-7