1-Bromoperfluorodecane 98%

Supplier: Apollo Scientific
Warning

PC450264-1G PC450264-25G PC450264-5G
APOSPC450264-1GEA 79.9 EUR
APOSPC450264-1G APOSPC450264-25G APOSPC450264-5G
1-Bromoperfluorodecane 98%
1-Bromoperfluorodecane
Formula: BrF₂C(CF₂)₈CF₃
Melting Pt: 55 °C
Density: 1.862 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 307-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR