1-(2-Bromoethyl)-3-(trifluoromethyl)benzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-2-[3-(trifluoromethyl)phenyl]ethane
PC450278-1G PC450278-5G
APOSPC450278-1GEA 78.8 EUR
APOSPC450278-1G APOSPC450278-5G
1-(2-Bromoethyl)-3-(trifluoromethyl)benzene 97%
1-(2-Bromoethyl)-3-(trifluoromethyl)benzene
Formula: C₉H₈BrF₃
MW: 253.062 g/mol
Boiling Pt: 90…92 °C/12mmHg
Density: 1.5 g/cm³ (25 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD03092900
CAS Number: 1997-80-4
UN: 3082
ADR: 9+EHS III