Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate
Supplier: Apollo Scientific
Warning
Synonyms: Phosphoric Acid Tris(1,1,1,3,3,3-hexafluoro-2-propyl) Ester
PC450502-1G PC450502-5G
APOSPC450502-1GEA 115 EUR
APOSPC450502-1G APOSPC450502-5G
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate
Tris(1,1,1,3,3,3-hexafluoro-2-propyl)phosphate
Formula: C₉H₃F₁₈O₄P
MW: 548.06 g/mol
Boiling Pt: 72 °C/12mmHg
Melting Pt: 32 °C
Storage Temperature: Ambient
CAS Number: 66489-68-7