2-[1-Methyl-5-(trifluoromethyl)pyrazol-3-yl]-thiophene-5-carboxaldehyde 97%
Supplier: Apollo Scientific
Warning
PC4518-1G PC4518-5G
APOSPC4518-1GEA 135 EUR
APOSPC4518-1G APOSPC4518-5G
2-[1-Methyl-5-(trifluoromethyl)pyrazol-3-yl]-thiophene-5-carboxaldehyde 97%
2-[1-Methyl-5-(trifluoromethyl)pyrazol-3-yl]thiophene-5-carboxaldehyde
Formula: C₁₀H₇F₃N₂OS
MW: 260.24 g/mol
Melting Pt: 80…81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052163
CAS Number: 175202-93-4