1H,1H,9H-Hexadecafluoro-1-nonanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1-nonanol, 1H,1H,9H-Perfluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluor-1-nonanol, 1H,1h,9h-hexadecafluorononanol
PC4530-100G PC4530-250G PC4530-25G
APOSPC4530-100GEA 92.8 EUR
APOSPC4530-100G APOSPC4530-250G APOSPC4530-25G
1H,1H,9H-Hexadecafluoro-1-nonanol 97%
1H,1H,9H-Hexadecafluoro-1-nonanol
Contains upto 10% H(CF2CF2)nCHOH(CF2CF2)mH where m+n=4.
Formula: C₉H₄F₁₆O
MW: 432.1 g/mol
Boiling Pt: 155…158 °C/200mmHg
Melting Pt: 53…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00039629
CAS Number: 376-18-1
EINECS: 206-806-0