1-(3-Fluorophenyl)-2-methylsulphonylethanone 95+%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3'-Fluoro-2-(methylsulphonyl)acetophenone
PC4541-1G
PC4541-5G
PC4541-25G
APOSPC4541-1GEA
72.5
EUR
APOSPC4541-1G
APOSPC4541-5G
APOSPC4541-25G
1-(3-Fluorophenyl)-2-methylsulphonylethanone 95+%
1-(3-Fluorophenyl)-2-methylsulphonylethanone
Formula:
C₉H₉FO₃S MW: 216.23 g/mol Melting Pt: 81…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD01941312 CAS Number: 82652-12-8 |