Hexafluoroacetone trihydrate 95%
Supplier: Apollo Scientific
Danger
Synonyms: Perfluoroacetone trihydrate
PC4571-100G PC4571-25G
APOSPC4571-100GEA 132 EUR
APOSPC4571-100G APOSPC4571-25G
Hexafluoroacetone trihydrate 95%
Hexafluoroacetone trihydrate
Formula: C₃F₆O·3H₂O
MW: 220.07 g/mol
Boiling Pt: 105…108 °C
Melting Pt: 18…21 °C
Density: 1.579 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149061
CAS Number: 34202-69-2
EINECS: 211-676-3
UN: 2552
ADR: 6.1 II