2,2,4,4-Tetrafluoro-1,3-dithietane
Supplier: Apollo Scientific
Warning
PC4572-100G PC4572-25G
APOSPC4572-100GEA 0 EUR
APOSPC4572-100G APOSPC4572-25G
2,2,4,4-Tetrafluoro-1,3-dithietane
2,2,4,4-Tetrafluoro-1,3-dithietane
Formula: C₂F₄S₂
MW: 164.14 g/mol
Boiling Pt: 47…50 °C
Density: 1.582 g/cm³ (25 °C)
Flash Pt: 35 °C
Storage Temperature: Ambient
MDL Number: MFCD00053538
CAS Number: 1717-50-6
EINECS: 217-001-9
UN: 1993
ADR: 3,III