Hexafluoroantimonic acid hexahydrate 98%
Supplier: Apollo Scientific
Warning
PC4593-100G PC4593-25G
APOSPC4593-100GEA 158 EUR
APOSPC4593-100G APOSPC4593-25G
Hexafluoroantimonic acid hexahydrate 98%
Hexafluoroantimonic acid hexahydrate
Formula: HF₆Sb·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149905
CAS Number: 72121-43-8
EINECS: 241-023-8
UN: 2922
ADR: 8+6.1,II