3-Chloro-5-fluoroaniline 98%
Supplier: Apollo Scientific
Warning
PC4608-5G PC4608-1G PC4608-25G
APOSPC4608-5GEA 34.8 EUR
APOSPC4608-5G APOSPC4608-1G APOSPC4608-25G
3-Chloro-5-fluoroaniline 98%
3-Chloro-5-fluoroaniline
Formula: C₆H₅ClFN
Density: 1.45 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD03407962
CAS Number: 4863-91-6
UN: 2810
ADR: 6.1,III