2-Amino-6-(6-fluoro-2-pyridinyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile 97%

Supplier: Apollo Scientific
Warning

PC4614-250MG PC4614-100MG
APOSPC4614-250MGEA 499 EUR
APOSPC4614-250MG APOSPC4614-100MG
2-Amino-6-(6-fluoro-2-pyridinyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile 97%
2-Amino-6-(6-fluoro-2-pyridinyl)-4,5,6,7-tetrahydrothieno[2,3-c]pyridine-3-carbonitrile
Formula: C₁₃H₁₁FN₄S
MW: 274.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00794643
CAS Number: 338413-86-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR