4-Amino-2,5-difluorobenzonitrile 98%
Supplier: Apollo Scientific
Warning
PC4621-1G PC4621-5G PC4621-25G PC4621-100G
APOSPC4621-1GEA 0 EUR
APOSPC4621-1G APOSPC4621-5G APOSPC4621-25G APOSPC4621-100G
4-Amino-2,5-difluorobenzonitrile 98%
4-Amino-2,5-difluorobenzonitrile
Formula: C₇H₄F₂N₂
MW: 154.12 g/mol
Melting Pt: 91…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00190102
CAS Number: 112279-61-5
UN: 3439
ADR: 6.1 III