Perfluorooctane 99%
Supplier: Apollo Scientific
Warning
Synonyms: Octadecafluorooctane
PC4640-25G PC4640-5G
APOSPC4640-25GEA 118 EUR
APOSPC4640-25G APOSPC4640-5G
Perfluorooctane 99%
Perfluorooctane
Formula: C₈F₁₈
MW: 438.06 g/mol
Boiling Pt: 99…105 °C
Melting Pt: -25
Density: 1.73 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042083
CAS Number: 307-34-6
EINECS: 206-199-2