2-(Heptafluoro-n-propyl)benzimidazole
Supplier: Apollo Scientific
Warning
PC4667-1G PC4667-5G
APOSPC4667-1GEA 94.8 EUR
APOSPC4667-1G APOSPC4667-5G
2-(Heptafluoro-n-propyl)benzimidazole
2-(Heptafluoro-n-propyl)benzimidazole
Formula: C₁₀H₅F₇N₂
MW: 286.15 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00521800
CAS Number: 559-37-5