2-Chloro-4-(trifluoromethyl)pyrimidine 97%
Supplier: Apollo Scientific
Warning
PC4691-100G PC4691-10G PC4691-25G
APOSPC4691-100GEA 149 EUR
APOSPC4691-100G APOSPC4691-10G APOSPC4691-25G
2-Chloro-4-(trifluoromethyl)pyrimidine 97%
2-Chloro-4-(trifluoromethyl)pyrimidine
Formula: C₅H₂ClF₃N₂
MW: 182.5310 g/mol
Boiling Pt: 60 °C/10mmHg
Density: 1.513 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00115111
CAS Number: 33034-67-2
UN: 1993
ADR: 3,III