1,4-Dibromohexafluoro-2-butene 97%
Supplier: Apollo Scientific
PC4718-25G PC4718-5G
APOSPC4718-25GEA 478 EUR
APOSPC4718-25G APOSPC4718-5G
1,4-Dibromohexafluoro-2-butene 97%
1,4-Dibromohexafluoro-2-butene
Formula: C₄Br₂F₆
MW: 321.85 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04038279
CAS Number: 360-87-2