2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 97%
Supplier: Apollo Scientific
Warning
APOSPC4740-1GEA 14.7 EUR
APOSPC4740-1G APOSPC4740-10G APOSPC4740-5G
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol 97%
2-(4-Aminophenyl)-1,1,1,3,3,3-hexafluoro-2-propanol
Formula: C₉H₇F₆NO
MW: 259,151 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039258
CAS Number: 722-92-9