1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1,1,1,3,3,3-Hexafluoro-2-methylpropan-2-ol, Hexafluoro-tert-butanol, Hexafluoro-2-methylisopropanol
PC4765-100G PC4765-25G PC4765-5G
APOSPC4765-100GEA 619 EUR
APOSPC4765-100G APOSPC4765-25G APOSPC4765-5G
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol 98%
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Formula: C₄H₄F₆O
Boiling Pt: 60…62 °C
Density: 1.484 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00039259
CAS Number: 1515-14-6