1,1,2,3,3,3-Hexafluoropropyl ethyl ether 97%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 1,1,2,3,3,3-Hexafluoropropyl ether
PC4795-25G PC4795-5G PC4795-1G
APOSPC4795-25GEA 174 EUR
APOSPC4795-25G APOSPC4795-5G APOSPC4795-1G
1,1,2,3,3,3-Hexafluoropropyl ethyl ether 97%
1,1,2,3,3,3-Hexafluoropropyl ethyl ether
Formula: C₅H₆F₆O
Boiling Pt: 64…71 °C
Density: 1.321 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236122
CAS Number: 380-34-7
UN: 3271
ADR: 3 II