2'-Trifluoromethoxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-[2-(Trifluoromethoxy)phenyl]-1-ethanone, 1-(2-(Trifluoromethoxy)phenyl)ethanone
PC4816-1G PC4816-25G PC4816-5G
APOSPC4816-1GEA 21.9 EUR
APOSPC4816-1G APOSPC4816-25G APOSPC4816-5G
2'-Trifluoromethoxyacetophenone 98%
2'-Trifluoromethoxyacetophenone
Formula: C₉H₇F₃O₂
MW: 204.15
Density: 1.265 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236320
CAS Number: 220227-93-0