4-Fluoro-3-methoxybenzoic acid 96%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methoxy-4-fluorobenzoic acid
PC4823-25G PC4823-5G PC4823-100G
APOSPC4823-25GEA 108 EUR
APOSPC4823-25G APOSPC4823-5G APOSPC4823-100G
4-Fluoro-3-methoxybenzoic acid 96%
4-Fluoro-3-methoxybenzoic acid
Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 206…208 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00272134
CAS Number: 82846-18-2